Methyl-3-(bromomethyl)benzoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Methyl 3-(bromomethyl)benzoate
M-Clarity™ quality level = MQ200
648116-25G 648116-5G
SIAL648116-25GEA 208 EUR
SIAL648116-25G SIAL648116-5G
Methyl-3-(bromomethyl)benzoate, Sigma-Aldrich®
Methyl 3-(bromomethyl)benzoate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD00051437
CAS Number: 1129-28-8
EINECS: 419-100-4