Methyl-4-(bromomethyl)benzoate

Supplier: Thermo Scientific

Synonyms: Methyl 4-(bromomethyl)benzoate

SEW03798.1 SEW03798EA SEW03798.50
MAYBSEW03798.1EA 0 EUR
MAYBSEW03798.1 MAYBSEW03798EA MAYBSEW03798.50
Methyl-4-(bromomethyl)benzoate
Methyl 4-(bromomethyl)benzoate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD00032453
CAS Number: 2417-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR