Climate chambers, ICH series
Supplier: MEMMERT
ICH110 ICH110L ICH260 ICH260L ICH260LT01 ICH260LT81 ICH750 ICH750L
390-0663EA 9969 EUR
390-0663 390-0665 390-0666 390-0668 MEMMICH260LT01 MEMMICH260LT81 390-0669 390-0671
Climate chambers, ICH series
Testing Chambers Photostability Chambers
The ICH range of climatic chambers offers rapid and precise temperature control from 10 up to 60 °C with humidification from 10 to 80% RH. These compressor cooled units have stainless steel inner chamber and housing, ensuring long working life and easy cleaning. The ICH models have humidity control, ICH L models feature humidity control and light and ICH C models have humidity and an electronic CO₂ control with automatic zero setting, NDIR measurement system and auto-diagnostic system.
  • Intuitive, user friendly TwinDISPLAY with 2 TFT displays.
More Product Information
Order Now
SPECIFICATIONS
Model ICH110 ICH110L ICH260 ICH260L ICH260L ICH750 ICH750L
Environmentally Preferable
Convection type Air jacket system Air jacket system Air jacket system Air jacket system Air jacket system Air jacket system Air jacket system
Humidity range 10...80% RH 10...80% RH 10...80% RH 10...80% RH 10...80% RH 10...80% RH
Temperature range 10...60 °C (ICH/ICH L with humidity and light, ICH C with humidity and CO₂)
0...60 °C (ICH L/C without humidity)
−10...+60 °C (ICH without humidity)
10...60 °C (ICH/ICH L with humidity and light, ICH C with humidity and CO₂)
0...60 °C (ICH L/C without humidity)
−10...+60 °C (ICH without humidity)
10...60 °C (ICH/ICH L with humidity and light, ICH C with humidity and CO₂)
0...60 °C (ICH L/C without humidity)
−10...+60 °C (ICH without humidity)
10...60 °C (ICH/ICH L with humidity and light, ICH C with humidity and CO₂)
0...60 °C (ICH L/C without humidity)
−10...+60 °C (ICH without humidity)
10...60 °C (ICH/ICH L with humidity and light, ICH C with humidity and CO₂)
0...60 °C (ICH L/C without humidity)
−10...+60 °C (ICH without humidity)
10...60 °C (ICH/ICH L with humidity and light, ICH C with humidity and CO₂)
0...60 °C (ICH L/C without humidity)
−10...+60 °C (ICH without humidity)
10...60 °C (ICH/ICH L with humidity and light, ICH C with humidity and CO₂)
0...60 °C (ICH L/C without humidity)
−10...+60 °C (ICH without humidity)
Temperature variation (spatial) ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C
Temperature fluctuation (time) ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C ≤±0,3 °C @ 40 °C