Anti-Myelin Protein Zero Rabbit Monoclonal Antibody [clone: EPR20383]

Supplier: Abcam
AB183868-100 AB183868-1ML
ABCAAB183868-100EA 732 EUR
ABCAAB183868-100 ABCAAB183868-1ML
Anti-Myelin Protein Zero Rabbit Monoclonal Antibody [clone: EPR20383]
Antibodies
Rabbit monoclonal [EPR20383] to Myelin Protein Zero.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Myelin Protein Zero
Clonality: Monoclonal
Clone: EPR20383
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR