Anti-CD26L Rabbit Monoclonal Antibody [clone: EPR18391-234]
Supplier: Abcam
AB305120-1 AB305120-100
ABCAAB305120-1EA 8710 EUR
ABCAAB305120-1 ABCAAB305120-100
Anti-CD26L Rabbit Monoclonal Antibody [clone: EPR18391-234]
Antibodies
Rabbit monoclonal [EPR18391-234] to CD26L - BSA and Azide free (Capture).
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD26L
Clonality: Monoclonal
Clone: EPR18391-234
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human