Anti-CD33 Rabbit Monoclonal Antibody [clone: EPR23052-20]
Supplier: Abcam
AB281179-1 AB281179-100
ABCAAB281179-1EA 8680 EUR
ABCAAB281179-1 ABCAAB281179-100
Anti-CD33 Rabbit Monoclonal Antibody [clone: EPR23052-20]
Antibodies
Rabbit monoclonal [EPR23052-20] to CD33 - BSA and Azide free (Capture).
Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: EPR23052-20
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human