Anti-EpCAM Rabbit Monoclonal Antibody [clone: EPR20532-222] (Alexa Fluor® 488)
Supplier: Abcam
AB313667-10UL AB313667-100UL
ABCAAB313667-10ULEA 238 EUR
ABCAAB313667-10UL ABCAAB313667-100UL
Anti-EpCAM Rabbit Monoclonal Antibody [clone: EPR20532-222] (Alexa Fluor® 488)
Antibodies
Alexa Fluor® 488 Rabbit monoclonal [EPR20532-222] to EpCAM.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: EpCAM
Clonality: Monoclonal
Clone: EPR20532-222
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat