Anti-Retinoic Acid Receptor beta Rabbit Monoclonal Antibody [clone: EPR2017]

Supplier: Abcam
AB247968-100 AB247968-1
ABCAAB247968-100EA 1020 EUR
ABCAAB247968-100 ABCAAB247968-1
Anti-Retinoic Acid Receptor beta Rabbit Monoclonal Antibody [clone: EPR2017]
Antibodies
Rabbit monoclonal [EPR2017] to Retinoic Acid Receptor beta - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Retinoic Acid Receptor beta
Clonality: Monoclonal
Clone: EPR2017
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR