Anti-ELMO1+ELMO2 Rabbit Monoclonal Antibody [clone: EPR13567]

Supplier: Abcam
AB181234-100 AB181234-1ML
ABCAAB181234-100EA 625 EUR
ABCAAB181234-100 ABCAAB181234-1ML
Anti-ELMO1+ELMO2 Rabbit Monoclonal Antibody [clone: EPR13567]
Antibodies
Rabbit monoclonal [EPR13567] to ELMO1 + ELMO2.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ELMO1+ELMO2
Clonality: Monoclonal
Clone: EPR13567
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR