Anti-Semaphorin 3A Rabbit Monoclonal Antibody [clone: EPR19367]

Supplier: Abcam
AB222497-100 AB222497-1
ABCAAB222497-100EA 1020 EUR
ABCAAB222497-100 ABCAAB222497-1
Anti-Semaphorin 3A Rabbit Monoclonal Antibody [clone: EPR19367]
Antibodies
Rabbit monoclonal [EPR19367] to Semaphorin 3A - Low endotoxin, Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Semaphorin 3A
Clonality: Monoclonal
Clone: EPR19367
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR