Anti-Glutamine Synthetase Rabbit Monoclonal Antibody [clone: EPR13022(B)] (Alexa Fluor® 568)
Supplier: Abcam
AB302807-100 AB302807-10
ABCAAB302807-100EA 832 EUR
ABCAAB302807-100 ABCAAB302807-10
Anti-Glutamine Synthetase Rabbit Monoclonal Antibody [clone: EPR13022(B)] (Alexa Fluor® 568)
Antibodies
Alexa Fluor® 568 Rabbit monoclonal [EPR13022(B)] to Glutamine Synthetase.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Glutamine Synthetase
Clonality: Monoclonal
Clone: EPR13022(B)
Conjugation: Alexa Fluor® 568
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human