Anti-CD4 Rabbit Monoclonal Antibody [clone: EPR19514]
Supplier: Abcam
AB221775-100 AB221775-1
ABCAAB221775-100EA 1020 EUR
ABCAAB221775-100 ABCAAB221775-1
Anti-CD4 Rabbit Monoclonal Antibody [clone: EPR19514]
Antibodies
Rabbit monoclonal [EPR19514] to CD4 - Low endotoxin, Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: EPR19514
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse