Anti-Eph receptor B2 Rabbit Monoclonal Antibody [clone: EPR22422-67]

Supplier: Abcam
AB307811-100 AB307811-20UL AB307811-1ML
ABCAAB307811-100EA 694 EUR
ABCAAB307811-100 ABCAAB307811-20UL ABCAAB307811-1ML
Anti-Eph receptor B2 Rabbit Monoclonal Antibody [clone: EPR22422-67]
Antibodies
Rabbit monoclonal [EPR22422-67] to Eph receptor B2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Eph receptor B2
Clonality: Monoclonal
Clone: EPR22422-67
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR