Anti-NFE2L1 Rabbit Monoclonal Antibody [clone: EPR25225-16]
Supplier: Abcam
AB302520-100 AB302520-20UL AB302520-1ML
ABCAAB302520-100EA 675 EUR
ABCAAB302520-100 ABCAAB302520-20UL ABCAAB302520-1ML
Anti-NFE2L1 Rabbit Monoclonal Antibody [clone: EPR25225-16]
Antibodies
Rabbit monoclonal [EPR25225-16] to NFE2L1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: NFE2L1
Clonality: Monoclonal
Clone: EPR25225-16
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat