Anti-Cytochrome P450 1A2 Rabbit Monoclonal Antibody [clone: EPR28416-16]

Supplier: Abcam
AB309542-100UL AB309542-20UL AB309542-1ML
ABCAAB309542-100ULEA 694 EUR
ABCAAB309542-100UL ABCAAB309542-20UL ABCAAB309542-1ML
Anti-Cytochrome P450 1A2 Rabbit Monoclonal Antibody [clone: EPR28416-16]
Antibodies
Rabbit monoclonal [EPR28416-16] to Cytochrome P450 1A2.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cytochrome P450 1A2
Clonality: Monoclonal
Clone: EPR28416-16
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR