Anti-Human Papillomavirus 16 (E7) Rabbit Monoclonal Antibody [clone: EPR25814-116]

Supplier: Abcam
AB300762-1 AB300762-100
ABCAAB300762-1EA 8960 EUR
ABCAAB300762-1 ABCAAB300762-100
Anti-Human Papillomavirus 16 (E7) Rabbit Monoclonal Antibody [clone: EPR25814-116]
Antibodies
Rabbit monoclonal [EPR25814-116] to Human Papillomavirus 16 (E7) - BSA and Azide free (Capture).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Human Papillomavirus 16 (E7)
Clonality: Monoclonal
Clone: EPR25814-116
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human papilloma virus (HPV)-16
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR