Anti-CD36 Rabbit Monoclonal Antibody [clone: EPR6573]

Supplier: Abcam
AB221605-100 AB221605-1
ABCAAB221605-100EA 1020 EUR
ABCAAB221605-100 ABCAAB221605-1
Anti-CD36 Rabbit Monoclonal Antibody [clone: EPR6573]
Antibodies
Rabbit monoclonal [EPR6573] to CD36 - Low endotoxin, Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: EPR6573
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR