Anti-SARS-CoV-2 Spike Glycoprotein S1 Rat Monoclonal Antibody [clone: CR3022]

Supplier: Abcam
AB281313-100 AB281313-1
ABCAAB281313-100EA 1020 EUR
ABCAAB281313-100 ABCAAB281313-1
Anti-SARS-CoV-2 Spike Glycoprotein S1 Rat Monoclonal Antibody [clone: CR3022]
Antibodies
Rat monoclonal [CR3022] to SARS-CoV-2 Spike Glycoprotein S1 - Chimeric – BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: SARS-CoV-2 Spike Glycoprotein S1
Clonality: Monoclonal
Clone: CR3022
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR