Anti-Dnmt3a Rabbit Monoclonal Antibody [clone: EPR24337-44]

Supplier: Abcam
AB271004-100 AB271004-20UL AB271004-1ML
ABCAAB271004-100EA 713 EUR
ABCAAB271004-100 ABCAAB271004-20UL ABCAAB271004-1ML
Anti-Dnmt3a Rabbit Monoclonal Antibody [clone: EPR24337-44]
Antibodies
Rabbit monoclonal [EPR24337-44] to Dnmt3a (mutated R882H).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Dnmt3a
Clonality: Monoclonal
Clone: EPR24337-44
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR