Anti-Fumarylacetoacetate hydrolase/FAA Rabbit Polyclonal Antibody

Supplier: Abcam
AB151998-100 AB151998-25
ABCAAB151998-100EA 688 EUR
ABCAAB151998-100 ABCAAB151998-25
Anti-Fumarylacetoacetate hydrolase/FAA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Fumarylacetoacetate hydrolase/FAA.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Fumarylacetoacetate hydrolase/FAA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR