Anti-Glycerol 3 Phosphate Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR25996-4]
Supplier: Abcam
AB313872-1MG AB313872-100UG
ABCAAB313872-1MGEA 9160 EUR
ABCAAB313872-1MG ABCAAB313872-100UG
Anti-Glycerol 3 Phosphate Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR25996-4]
Antibodies
Rabbit monoclonal [EPR25996-4] to Glycerol 3 Phosphate Dehydrogenase - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Glycerol 3 Phosphate Dehydrogenase
Clonality: Monoclonal
Clone: EPR25996-4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat