Anti-Ctip1/BCL-11A Mouse Monoclonal Antibody [clone: 123]

Supplier: Abcam
AB256127-1 AB256127-100
ABCAAB256127-1EA 9160 EUR
ABCAAB256127-1 ABCAAB256127-100
Anti-Ctip1/BCL-11A Mouse Monoclonal Antibody [clone: 123]
Antibodies
Mouse monoclonal [123] to Ctip1/BCL-11A - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Ctip1/BCL-11A
Clonality: Monoclonal
Clone: 123
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR