Anti-HIV1 p55 + p17 Rabbit Monoclonal Antibody [clone: EPR27327-41]

Supplier: Abcam
AB308070-100 AB308070-20UL AB308070-1ML
ABCAAB308070-100EA 694 EUR
ABCAAB308070-100 ABCAAB308070-20UL ABCAAB308070-1ML
Anti-HIV1 p55 + p17 Rabbit Monoclonal Antibody [clone: EPR27327-41]
Antibodies
Rabbit monoclonal [EPR27327-41] to HIV1 p55 + p17.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HIV1 p55 + p17
Clonality: Monoclonal
Clone: EPR27327-41
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: HIV
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR