Anti-ProDynorphin Rabbit Monoclonal Antibody [clone: EPR24508-78]
Supplier: Abcam
AB283501-100 AB283501-20UL AB283501-1ML
ABCAAB283501-100EA 713 EUR
ABCAAB283501-100 ABCAAB283501-20UL ABCAAB283501-1ML
Anti-ProDynorphin Rabbit Monoclonal Antibody [clone: EPR24508-78]
Antibodies
Rabbit monoclonal [EPR24508-78] to ProDynorphin.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ProDynorphin
Clonality: Monoclonal
Clone: EPR24508-78
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat