Anti-Rab9 Rabbit Monoclonal Antibody [clone: EPR13272]

Supplier: Abcam
AB179815-100 AB179815-1ML
ABCAAB179815-100EA 475 EUR
ABCAAB179815-100 ABCAAB179815-1ML
Anti-Rab9 Rabbit Monoclonal Antibody [clone: EPR13272]
Antibodies
Rabbit monoclonal [EPR13272] to Rab9 - Late Endosome Marker.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Rab9
Clonality: Monoclonal
Clone: EPR13272
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR