Anti-Cytokeratin 17 Rabbit Monoclonal Antibody [clone: EPR20300-48]

Supplier: Abcam
AB259570-100 AB259570-1
ABCAAB259570-100EA 938 EUR
ABCAAB259570-100 ABCAAB259570-1
Anti-Cytokeratin 17 Rabbit Monoclonal Antibody [clone: EPR20300-48]
Antibodies
Rabbit monoclonal [EPR20300-48] to Cytokeratin 17 - BSA and Azide free (Detector).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cytokeratin 17
Clonality: Monoclonal
Clone: EPR20300-48
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR