Anti-pan-myc Rabbit Monoclonal Antibody [clone: EPR18863]

Supplier: Abcam
AB195207-100 AB195207-20UL AB195207-1ML
ABCAAB195207-100EA 569 EUR
ABCAAB195207-100 ABCAAB195207-20UL ABCAAB195207-1ML
Anti-pan-myc Rabbit Monoclonal Antibody [clone: EPR18863]
Antibodies
Rabbit monoclonal [EPR18863] to pan-myc.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: pan-myc
Clonality: Monoclonal
Clone: EPR18863
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR