Anti-ALPK1 Rabbit Monoclonal Antibody [clone: EPR20911-169]

Supplier: Abcam
AB236626-100 AB236626-20UL AB236626-1ML
ABCAAB236626-100EA 713 EUR
ABCAAB236626-100 ABCAAB236626-20UL ABCAAB236626-1ML
Anti-ALPK1 Rabbit Monoclonal Antibody [clone: EPR20911-169]
Antibodies
Rabbit monoclonal [EPR20911-169] to ALPK1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ALPK1
Clonality: Monoclonal
Clone: EPR20911-169
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR