Anti-EIF2S1 Rabbit Monoclonal Antibody [clone: E90]

Supplier: Abcam
AB214434-100 AB214434-1
ABCAAB214434-100EA 1020 EUR
ABCAAB214434-100 ABCAAB214434-1
Anti-EIF2S1 Rabbit Monoclonal Antibody [clone: E90]
Antibodies
Rabbit monoclonal [E90] to EIF2S1 (phospho S51) - BSA and azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: EIF2S1
Clonality: Monoclonal
Clone: E90
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human, N cra, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR