Anti-Pan Trk Rabbit Monoclonal Antibody [clone: EPR24009-335]

Supplier: Abcam
AB267830-100 AB267830-20UL AB267830-1ML
ABCAAB267830-100EA 707 EUR
ABCAAB267830-100 ABCAAB267830-20UL ABCAAB267830-1ML
Anti-Pan Trk Rabbit Monoclonal Antibody [clone: EPR24009-335]
Antibodies
Rabbit monoclonal [EPR24009-335] to Pan Trk.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Pan Trk
Clonality: Monoclonal
Clone: EPR24009-335
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR