Anti-Sodium/potassium-transporting ATPase (pan alpha subunit) Rabbit Monoclonal Antibody [clone: EPR25414-249]

Supplier: Abcam
AB300507-100 AB300507-20UL AB300507-1ML
ABCAAB300507-100EA 707 EUR
ABCAAB300507-100 ABCAAB300507-20UL ABCAAB300507-1ML
Anti-Sodium/potassium-transporting ATPase (pan alpha subunit) Rabbit Monoclonal Antibody [clone: EPR25414-249]
Antibodies
Rabbit monoclonal [EPR25414-249] to Sodium/potassium-transporting ATPase (pan alpha subunit).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Sodium/potassium-transporting ATPase (pan alpha subunit)
Clonality: Monoclonal
Clone: EPR25414-249
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR