Anti-Scavenging Receptor SR-BI Rabbit Monoclonal Antibody [clone: EPR20190]

Supplier: Abcam
AB222931-100 AB222931-1
ABCAAB222931-100EA 1020 EUR
ABCAAB222931-100 ABCAAB222931-1
Anti-Scavenging Receptor SR-BI Rabbit Monoclonal Antibody [clone: EPR20190]
Antibodies
Rabbit monoclonal [EPR20190] to Scavenging Receptor SR-BI - Low endotoxin, Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Scavenging Receptor SR-BI
Clonality: Monoclonal
Clone: EPR20190
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR