Anti-MSH2 Rabbit Monoclonal Antibody [clone: EPR21017-123]

Supplier: Abcam
AB227941-100 AB227941-1ML
ABCAAB227941-100EA 657 EUR
ABCAAB227941-100 ABCAAB227941-1ML
Anti-MSH2 Rabbit Monoclonal Antibody [clone: EPR21017-123]
Antibodies
Rabbit monoclonal [EPR21017-123] to MSH2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MSH2
Clonality: Monoclonal
Clone: EPR21017-123
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR