Anti-GCDFP 15 Rabbit Monoclonal Antibody [clone: SP185]
Supplier: Abcam
AB183321-100 AB183321-500 AB183321-1
ABCAAB183321-100EA 688 EUR
ABCAAB183321-100 ABCAAB183321-500 ABCAAB183321-1
Anti-GCDFP 15 Rabbit Monoclonal Antibody [clone: SP185]
Antibodies
Rabbit monoclonal [SP185] to GCDFP 15 - C-terminal.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GCDFP 15
Clonality: Monoclonal
Clone: SP185
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human