Anti-NPAT Mouse Monoclonal Antibody [clone: 27/NPAT]
Supplier: Abcam
AB307837-100 AB307837-10 AB307837-20UG AB307837-1MG
ABCAAB307837-100EA 694 EUR
ABCAAB307837-100 ABCAAB307837-10 ABCAAB307837-20UG ABCAAB307837-1MG
Anti-NPAT Mouse Monoclonal Antibody [clone: 27/NPAT]
Antibodies
Mouse monoclonal [27/NPAT] to NPAT.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: NPAT
Clonality: Monoclonal
Clone: 27/NPAT
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human