Anti-IgG Goat Polyclonal Antibody (HRP polymer)

Supplier: Abcam
AB214882-15 AB214882-50
ABCAAB214882-15EA 407 EUR
ABCAAB214882-15 ABCAAB214882-50
Anti-IgG Goat Polyclonal Antibody (HRP polymer)
Antibodies
Goat Anti-Rat IgG H&L (HRP polymer).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP polymer
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR