Anti-Sodium/calcium exchanger 2 Rabbit Monoclonal Antibody [clone: EPR25106-37]
Supplier: Abcam
AB300455-100 AB300455-20UL AB300455-1ML
ABCAAB300455-100EA 707 EUR
ABCAAB300455-100 ABCAAB300455-20UL ABCAAB300455-1ML
Anti-Sodium/calcium exchanger 2 Rabbit Monoclonal Antibody [clone: EPR25106-37]
Rabbit monoclonal [EPR25106-37] to Sodium/calcium exchanger 2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Sodium/calcium exchanger 2
Clonality: Monoclonal
Clone: EPR25106-37
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat