Anti-CD8 beta Rabbit Monoclonal Antibody [clone: EPR26538-16]
Supplier: Abcam
AB300067-100 AB300067-20UL AB300067-1ML
ABCAAB300067-100EA 707 EUR
ABCAAB300067-100 ABCAAB300067-20UL ABCAAB300067-1ML
Anti-CD8 beta Rabbit Monoclonal Antibody [clone: EPR26538-16]
Antibodies
Rabbit monoclonal [EPR26538-16] to CD8 beta.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD8 beta
Clonality: Monoclonal
Clone: EPR26538-16
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human