Anti-IgG Goat Polyclonal Antibody (HRP polymer)

Supplier: Abcam
AB214880-15 AB214880-50
ABCAAB214880-15EA 413 EUR
ABCAAB214880-15 ABCAAB214880-50
Anti-IgG Goat Polyclonal Antibody (HRP polymer)
Antibodies
Goat Anti-Rabbit IgG H&L (HRP polymer).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase) polymer
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR