Anti-CD147 Rabbit Monoclonal Antibody [clone: EPR18008-67]

Supplier: Abcam
AB212057-100 AB212057-1ML
ABCAAB212057-100EA 688 EUR
ABCAAB212057-100 ABCAAB212057-1ML
Anti-CD147 Rabbit Monoclonal Antibody [clone: EPR18008-67]
Antibodies
Rabbit monoclonal [EPR18008-67] to CD147.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD147
Clonality: Monoclonal
Clone: EPR18008-67
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR