Anti-MTTP/MTP Rabbit Monoclonal Antibody [clone: EPR27197-78]
Supplier: Abcam
AB321881-20UL AB321881-100UL AB321881-1ML
ABCAAB321881-20ULEA 244 EUR
ABCAAB321881-20UL ABCAAB321881-100UL ABCAAB321881-1ML
Anti-MTTP/MTP Rabbit Monoclonal Antibody [clone: EPR27197-78]
Antibodies
Rabbit monoclonal [EPR27197-78] to MTTP/MTP.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: MTTP/MTP
Clonality: Monoclonal
Clone: EPR27197-78
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse