Anti-Hepatitis B Virus Surface Antigen Mouse Monoclonal Antibody [clone: 3B8]

Supplier: Abcam
AB316775-1MG AB316775-100UG
ABCAAB316775-1MGEA 9160 EUR
ABCAAB316775-1MG ABCAAB316775-100UG
Anti-Hepatitis B Virus Surface Antigen Mouse Monoclonal Antibody [clone: 3B8]
Antibodies
Mouse monoclonal [3B8] to Hepatitis B Virus Surface Antigen - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Hepatitis B Virus Surface Antigen
Clonality: Monoclonal
Clone: 3B8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hepatitis B virus (HBV)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR