Anti-Total OXPHOS Antibody Cocktail Rabbit Recombinant Multiclonal Antibody [clone: RM1134]

Supplier: Abcam
AB317270-20UL AB317270-100UL AB317270-1ML
ABCAAB317270-20ULEA 207 EUR
ABCAAB317270-20UL ABCAAB317270-100UL ABCAAB317270-1ML
Anti-Total OXPHOS Antibody Cocktail Rabbit Recombinant Multiclonal Antibody [clone: RM1134]
Antibodies
Rabbit recombinant multiclonal [RM1134] to Total OXPHOS Antibody Cocktail.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Total OXPHOS Antibody Cocktail
Clonality: Recombinant Multiclonal
Clone: RM1134
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR