Anti-GPR56 Rabbit Monoclonal Antibody [clone: EPR26546-78] (Alexa Fluor® 647)

Supplier: Abcam
AB315851-10UL AB315851-100UL
ABCAAB315851-10ULEA 238 EUR
ABCAAB315851-10UL ABCAAB315851-100UL
Anti-GPR56 Rabbit Monoclonal Antibody [clone: EPR26546-78] (Alexa Fluor® 647)
Antibodies
Alexa Fluor® 647 Rabbit monoclonal [EPR26546-78] to GPR56.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: GPR56
Clonality: Monoclonal
Clone: EPR26546-78
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR