Power Protect V1000 Safety Gloves, 60840 Series
Supplier: UVEX
6084007 6084008 6084009 6084010 6084011
UVEX6084007EA 61 EUR
UVEX6084007 UVEX6084008 UVEX6084009 UVEX6084010 UVEX6084011
Power Protect V1000 Safety Gloves, 60840 Series
Gloves
The power protect V1000 protective glove provides reliable protection against electrical voltages up to 1000 V.
  • Natural latex protects the wearer from electrical discharges
  • Highly insulating
  • Excellent fit
  • Very flexible
Made from highly insulating natural latex, these gloves shield the wearer from electrical discharges. Additionally, their flexible design ensures comfortable wear and excellent tactile sensitivity.