Human TRAIL ELISA Kit (CD253)
Supplier: Abcam
AB46074-96 AB46074-2X96
ABCAAB46074-96EA 969 EUR
ABCAAB46074-96 ABCAAB46074-2X96
Human TRAIL ELISA Kit (CD253)
Assays ELISAs
Human TRAIL ELISA kit (CD253) is a Sandwich (quantitative) ELISA kit for the measurement of Human TRAIL (CD253) in Human in plasma, cell culture supernatant, serum samples.