Tris Buffered Saline (TBS) Casein ELISA Reagent
Supplier: Abcam
AB171533-100 AB171533-1000
ABCAAB171533-100EA 125 EUR
ABCAAB171533-100 ABCAAB171533-1000
Tris Buffered Saline (TBS) Casein ELISA Reagent
Assays ELISAs
Tris Buffered Saline (TBS) Casein ELISA Reagent is a buffer designed for ELISA assay development.