Human GM-CSF Receptor alpha ELISA Kits (CD116)
Supplier: Abcam
AB46086-96
AB46086-2X96
ABCAAB46086-96EA
900
EUR
ABCAAB46086-96
ABCAAB46086-2X96
Human GM-CSF Receptor alpha ELISA Kits (CD116)
Assays
ELISAs
Human GM-CSF Receptor alpha ELISA kit (CD116) is a Sandwich (quantitative) ELISA kit for the measurement of Human GM-CSF Receptor alpha (CD116) in Human in plasma, cell culture supernatant, Serum samples.