Human c-Kits ELISA Kits (CD117)
Supplier: Abcam
AB45924-96
AB45924-2X96
ABCAAB45924-96EA
969
EUR
ABCAAB45924-96
ABCAAB45924-2X96
Human c-Kits ELISA Kits (CD117)
Assays
ELISAs
Human c-kit ELISA kit (CD117) is a Sandwich (quantitative) ELISA kit for the measurement of Human c-kit (CD117) in Human in plasma, cell culture supernatant, Serum samples.