Human IL-17F ELISA Set (without plates)
Supplier: Abcam
AB105233-480 AB105233-10X96
ABCAAB105233-480EA 1370 EUR
ABCAAB105233-480 ABCAAB105233-10X96
Human IL-17F ELISA Set (without plates)
Assays ELISAs
Human IL-17F ELISA Set (without plates) is a sandwich (quantitative) ELISA kit for the measurement of Human IL-17F ELISA Set (without plates) in Plasma, cell culture supernatant, serum samples.